Learn More
Thermo Scientific™ CellSensor™ c-Fos-bla HEK293T Cell Line
The c-fos-bla HEK 293T cell line contains a beta-lactamase reporter gene under control of the c-fos promoter stably integrated into HEK 293T cells.
Brand: Thermo Scientific™ K1659B
Code : NEW
Additional Details : Weight : 0.50000kg
Description
This cell line can be used to detect agonists/antagonists of the c-fos pathway. The c-fos-bla HEK 293T cells have been shown to be responsive to phorbol 12-myristate 13-acetate (PMA) stimulation. Academic and non-profit customers, please inquire for special pricing.
Specifications
Dividing Cells | |
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. | |
CellSensor™ | |
Signaling Pathway |
For Research Use Only. Not for use in diagnostic procedures.