Learn More
Thermo Scientific™ CellSensor™ CRE-bla HEK 293T Cell Line
Description
The CellSensor™ CRE-bla HEK 293T cell line contains the b-lactamase reporter gene under control of the cAMP response element (CRE). To obtain the cell line, the CRE-bla construct was transduced into HEK 293T cells by lentivirus. Subsequent flow cytometry was used to isolate clones responsive to forskolin stimulation. The CellSensor™ CRE-bla HEK 293T cell line can be used to build specific G-protein coupled receptor (GPCR) assays or to detect changes in intracellular cAMP levels. Academic and non-profit customers, please inquire for special pricing.
Specifications
Specifications
| Assay Entry | Cell-based beta lactamase reporter gene |
| Cell State | Dividing Cells |
| Content And Storage | CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use. |
| Description | CellSensor™ CRE-bla HEK 293T Cell Line, 1Vial, ∽ 2,000,000 cells/vial at 2,000,000 cells/mL Quantity of Cells, CRE-bla HEK 293T Cell Line, Fluorescent Detection Method, CellSensor™ Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage |
| Detection Method | Fluorescent |
| For Use With (Application) | Pharma and biopharma, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, pathway biology, cellular pathway analysis assays, cellSensor™ cellular pathway assays |
| Includes | b-lactamase reporter gene under control of the cAMP response element (CRE) |
| Product Line | CellSensor |
| Quantity | 1 Vial |
| Sample Type | CRE-bla HEK 293T |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.