Learn More
Thermo Scientific™ CellSensor™ NFκB-bla RA 1 Cell Line
The CellSensor™ NFκB-bla RA-1 cell line contains a beta-lactamase reporter gene under control of the NFκB response element stably integrated into Ramos 1 (RA-1) cells.
Brand:  Thermo Scientific™ K1665
Product Code. 10638733
  • £7880.00 / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
The CellSensor™ NFκB-bla RA-1 cell line contains a beta-lactamase reporter gene under control of the NFκB response element stably integrated into Ramos 1 (RA-1) cells. To construct this cell line, the NFκB -bla construct was transduced into RA-1 cells by lentivirus. Flow cytometry was used to isolate cells responsive to Tumor Necrosis Factor (TNFThis cell line was validated for DMSO tolerance, cell number, stimulation time, substrate loading time, as well as Z' and EC50 concentrations for TNF. The CellSensor™ NFκB-bla RA-1 cell line is responsive to TNFand can be used to probe NFκB signaling pathways, which are involved in the regulation of apoptosis, viral replication, tumorigenesis, inflammation, and various autoimmune diseases. This cell line can be adapted for high-throughput screening for agonists or antagonists with compound libraries. Candidate drugs can be tested for dose response against this cell line. Academic and non-profit customers, please inquire for special pricing.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cell-based beta lactamase reporter gene
CellSensor™ Cell Lines are shipped on dry ice. Store in liquid nitrogen immediately upon receipt, or thaw for immediate use.
CellSensor™
TNF alpha, NIK, TLR, I kappa B, IRAK, IKK, NF-kappa B pathway
CellSensor™
Dividing Cells
Cytokines, Signaling Pathway
1 Vial
Dividing Cells
Signaling Pathway
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ CellSensor™ NFκB-bla RA 1 Cell Line > 1 Vial