Learn More
Invitrogen™ CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line
CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line
Brand: Invitrogen™ K1515
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Neurotrophins (NGF, BDNF, NT-3, and NT-4) and their transmembrane receptors (TrkA, TrkB, TrkC, and P75NTR) play important roles in the regulation of neuronal and non-neuronal cell proliferation, differentiation, survival, and death. Neurotrophin signaling also mediates higher-order neuronal activities, such as learning, memory, and behavior. Alterations in neurotrophin levels and their receptors have been implicated in neurodegenerative disorders such as Alzheimer's disease and Huntington's disease, as well as psychiatric disorders. Neurotrophins propagate their signal through activating multiple signaling pathways. One of the signaling pathways of NT-3 (the ligand for TrkC) activates phospholipase C, releasing DAG and IP3, increasing downstream intracellular calcium, and activating protein kinase C. This in turn promotes the translocation of the transcription factor-nuclear factor of activated T-cells (NFAT)-from the cytosol into the nucleus, resulting in NFAT-dependent transcription. The CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line was engineered by integrating the human TrkC expression plasmid into the genome of the CellSensor™ NFAT-bla CHO-K1 cell line, which is engineered to express beta-lactamase under the control of NFAT. This cell line has been tested for assay performance under variable conditions, including DMSO concentration, cell number, stimulation time, substrate loading time, and in cryopreserved cells, and has been validated for Z'-factor and EC50 values under optimized conditions using NT-3. Additional testing information using various small-molecule inhibitors and RNAi has been performed.Specifications
Liquid Nitrogen | |
Fluorescent | |
1 Vial | |
CHO-K1 Cells |
CellSensor™ TrkC-NFAT-bla CHO-K1 Cell Line, 1Vial, ∽ 8,000,000 cells/vial at 8,000,000 cells/mL Quantity of Cells, CHO-K1 Cell Line, Fluorescent Detection Method, Signaling Pathway Druggable Target, Adherent Growth Properties, Liquid Nitrogen Storage | |
Pharma and biopharma, drug discovery and development, target and lead identification and validation, kinase biology, kinase pathway analysis, pathway biology, cellular pathway analysis assays, cellSensor™ cellular pathway assays | |
CHO-K1 |
For Research Use Only. Not for use in diagnostic procedures.