missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One CELLSTAR 96-well, Cell Culture-Treated, Flat-Bottom Microplate

Color: Clear; Sterility: Sterile; Lid: Without Lid

Brand:  Greiner Bio-One 655160

 View more versions of this product

Product Code. 10430571

  • £173.00 / Pack of 100

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-Pyrogenic
  • Compatible with automated systems
  • Alphanumeric well coding
  • High clarity of microplates for optimal microscopic examinations
  • Stackable
  • Individu
    TRUSTED_SUSTAINABILITY
Specifications

Specifications

No
Without Lid
8 x 12
Clear
Polystyrene
25 to 340 μL
Cell Culture
Flat Bottom (F-well)
100
100/Cs.
Chimney
Standard
34 mm2
1/Pk., 100/Cs.
Tissue Culture Plate
96
Sterile
392 μL
4800 x g (swinging bucket rotor)
1
Clear
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers