Learn More
Greiner Bio-One CELLSTAR Cell Culture Flasks
Manufactured from high-grade polystyrene physically surface-treated to improve cell adhesion and proliferation. Greiner Bio-One™ CELLSTAR Cell Culture Flasks are designed to efficiently reach the cell lawn with a cell scraper and provide optimal access with a pipet.
Brand:  Greiner Bio-One 690160
 View more versions of this product
Product Code. 10150431
  • £297.00 / Pack of 200
Explore more special offers
Description
Description
  • Improved cell adhesion through physical surface treatment
  • Neck of high profile flask reduces the risk of wetting the inside of the cap with medium;  provides additional protection against contamination
  • Canted neck
  • TC surface treatment
  • Graduation on the side
  • Stackable
  • Measuring grid
  • Red standard screw cap
  • Sterile and user-friendly packaging
  • Free of detectable DNase, RNase, human DNA, non-pyrogenic
  • Non-Cytotoxic 
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Red
50 mL
Printed and engraved graduations
Physical Surface Treatment
Sterile
25 cm2
HDPE
Polystyrene
Canted
Yes
Screw Cap with Vent Position
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Greiner Bio-One CELLSTAR Cell Culture Flasks > Screw Cap Type: Red Filter Screw Cap; Capacity: 50mL; Stackable Flask