missing translation for 'onlineSavingsMsg'
Learn More

Biodesign™ Cellulose Dialysis Tubing Strips

Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.

Brand:  Biodesign™ D116

Code : LC

Additional Details : Weight : 0.20000kg

 View more versions of this product

Product Code. 12757486

  • £53.55 / Pack of 50
Estimated Shipment: 13-08-2024
to see stock.

Description

Description

  • Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
  • Just wet in distilled water and use
  • Available in 305mm strips
  • Specially purified for life science research
  • Supplied in an airtight, moisture-proof container for maximum shelf life
  • Available in 14000Da, 8000Da, and 3500Da molecular weight cut offs
Specifications

Specifications

Dialysis Tubing Strips
Cellulose
30.5 m
Transparent
For life sciences research
8 kDa
28.7 mm
6.45 mL/cm
1 Each
Product Suggestions

Product Suggestions

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales