Learn More
Biodesign™ Cellulose Dialysis Tubing Strips
Suitable for all types of laboratory dialysis including desalting, compound separation and binding studies.
£39.35 - £231.00
Specifications
Product Type | Dialysis Tubing Strips |
---|---|
Material | Cellulose |
Length (Metric) | 30.5 m |
Color | Transparent |
Quantity | 1 Each |
Product Code | Brand | Capacity (Metric) | MWCO | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | MWCO | Price | Quantity & Availability | ||||
12727476
|
Biodesign™
D011 |
1.32 mL/cm | 14 kDa |
£69.10
Pack of 50 |
Estimated Shipment: 24-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12737476
|
Biodesign™
D012 |
1.91 mL/cm | 14 kDa |
£54.60
Pack of 50 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12747476
|
Biodesign™
D014 |
3.57 mL/cm | 14 kDa |
£40.20
Pack of 50 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12757476
|
Biodesign™
D016 |
6.45 mL/cm | 14 kDa |
£117.00
Pack of 50 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12727486
|
Biodesign™
D110 |
0.32 mL/cm | 8 kDa |
£39.35
Pack of 50 |
Estimated Shipment: 11-11-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12737486
|
Biodesign™
D112 |
1.91 mL/cm | 8 kDa |
£53.45
Pack of 50 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12747486
|
Biodesign™
D114 |
3.57 mL/cm | 8 kDa |
£39.35
Pack of 50 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12757486
|
Biodesign™
D116 |
6.45 mL/cm | 8 kDa |
£53.55
Pack of 50 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12767486
|
Biodesign™
D118 |
19.13 mL/cm | 8 kDa |
£70.70
Pack of 50 |
Estimated Shipment: 24-10-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12707496
|
Biodesign™
D312 |
1.91 mL/cm | 3.5 kDa |
£231.00
Pack of 50 |
Estimated Shipment: 19-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12727496
|
Biodesign™
D316 |
6.45 mL/cm | 3.5 kDa |
£231.00
Pack of 50 |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
12717496
|
Biodesign™
D314 |
3.57 mL/cm | 3.5 kDa |
£88.70
Pack of 50 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Seamless, thin, transparent dialysis tubing manufactured from the highest purity cellulose
- Just wet in distilled water and use
- Available in 305mm strips
- Specially purified for life science research
- Supplied in an airtight, moisture-proof container for maximum shelf life
- Available in 14, 000Da, 8, 000Da, and 3, 500Da molecular weight cut offs
Specifications
Dialysis Tubing Strips | |
30.5 m | |
1 Each |
Cellulose | |
Transparent | |
For life sciences research |