missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Ceramus™ Fix Bottle Top Dispensers
Guarantees consistent precision and volume dispensing with reproducible results independent of the user. Hirschmann™ Ceramus™ Classic Dispensers are the proven unit for manual dispensing, featuring superior-quality materials, simple handling and well-engineered user comfort to make laboratory routines easier.
Brand: Hirschmann™ 9342100
This item is not returnable.
View return policy
Description
- For fixed and variable volumes, from 0.2mL to 60mL
- Complete unit can be steam sterilized at 121°C without dismantling
- Free rotation
- Media recirculating system for simple and rapid venting without reagent loss
- Digital volume configuration engagement, calibrated for a lifetime
- Use within a temperature range of 4°C to 50°C
- Individual quality certificate with conformity certificate
Specifications
Bottletop Dispenser | |
10 mL | |
0.6% |
Ceramic | |
Bottle |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction