missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hirschmann™ Ceramus™ Fix Bottle Top Dispensers
Guarantees consistent precision and volume dispensing with reproducible results independent of the user. Hirschmann™ Ceramus™ Classic Dispensers are the proven unit for manual dispensing, featuring superior-quality materials, simple handling and well-engineered user comfort to make laboratory routines easier.
£429.00 - £616.00
Specifications
Product Type | Bottletop Dispenser |
---|---|
Material | Ceramic |
For Use With (Equipment) | Bottle |
Precision | 0.6% |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | |||||
10548062
|
Hirschmann™
9362100 |
50 mL |
£616.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10267910
|
Hirschmann™
9342100 |
10 mL |
£429.00
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- For fixed and variable volumes, from 0.2mL to 60mL
- Complete unit can be steam sterilized at 121°C without dismantling
- Free rotation
- Media recirculating system for simple and rapid venting without reagent loss
- Digital volume configuration engagement, calibrated for a lifetime
- Use within a temperature range of 4°C to 50°C
- Individual quality certificate with conformity certificate
Specifications
Bottletop Dispenser | |
Ceramic | |
Bottle | |
0.6% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title