missing translation for 'onlineSavingsMsg'
Learn More

Gestigkeit Harry™ CERAN™ Hotplate, Glass Ceramic

Size: 10.2 x 10.2 in.

Brand:  Gestigkeit Harry™ 11EB-C

Product Code. 11784838

  • £1650.00 / Each
Estimated Shipment: 28-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Unusual glass ceramic material
  • Resistant to breakage and temperature changes
  • Free from distortion
  • Permeable to ultra-violet light and highly acid-resistant
Specifications

Specifications

Hot Plate
230 V
10.2 x 10.2 in.
Square
Glass Ceramic
2000 W
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gestigkeit Harry™ CERAN™ Hotplate, Glass Ceramic > Size: 10.2 x 10.2 in.

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.