missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauda™ Algizid Aquastab
Algizid Aquastab for Algae Control in Water Baths.
Brand: Lauda™ LZB 929
This item is not returnable.
View return policy
Description
- Highly effective algae control agent only for use with water in bath thermostats
- With this medium, the bacteriostatic and fungistatic stress of the water is kept low
Specifications
Blue, Clear | |
Composition: 2% Poly(dimethyl imino)-2-hydroxy-1,3-propanediyl chloride | |
100 mL | |
5°C to 90°C |
Product Suggestions
Customers who viewed this item also viewed.
Safety and Handling
Product Identifier
Hazard Category
Hazard Statement
Precautionary Statement
- Algizid Aquastab
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P102-Keep out of reach of children.
- P273-Avoid release to the environment.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Lauda™ Algizid Aquastab > Capacity: 100 mL
Spot an opportunity for improvement?Share a Content Correction