missing translation for 'onlineSavingsMsg'
Learn More

Cerium(III) chloride heptahydrate, 99%, Thermo Scientific Chemicals

£30.85 - £287.43

Chemical Identifiers

CAS 18618-55-8
Molecular Formula CeCl3·7H2O
MDL Number MFCD00149634
Synonym Cerous chloride heptahydrate
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11409793
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12947.18
50 g
£30.85
50g
Please to purchase this item. Need a web account? Register with us today!
11419793
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12947.30
250 g
£90.95
250g
Please to purchase this item. Need a web account? Register with us today!
11499783
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12947.0B
1000 g
£287.43
1000g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Cerium(III) chloride heptahydrate is used in the preparation of allylsilanes from esters. It is used as a reducing agent in organic synthesis in place of sodium borohydride. In Luche reaction, carvone gives selectively allylic alcohol.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cerium(III) chloride heptahydrate is used in the preparation of allylsilanes from esters. It is used as a reducing agent in organic synthesis in place of sodium borohydride. In Luche reaction, carvone gives selectively allylic alcohol.

Solubility
Soluble in alcohol and acetone. Slightly soluble in tetrahydrofuran. Insoluble in cold water.

Notes
Air sensitive. Hygroscopic. Incompatible with strong acids and strong oxidizing agents.
Specifications

Chemical Identifiers

18618-55-8
CeCl3·7H2O
MFCD00149634
Cerous chloride heptahydrate

Specifications

18618-55-8
MFCD00149634
Cerous chloride heptahydrate
372.60 (246.48 Anhydrous)
Odorless
Cerium(III) chloride heptahydrate
CeCl3·7H2O
14,1997
Soluble in alcohol,water,and acetone. Slightly soluble in tetrahydrofuran.
99%
Air sensitive; Hygroscopic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314

missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: II

missing translation for 'einecsNumber' : 606-073-0

missing translation for 'rtecsNumber' : FK5100000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Argon

Product Identifier
  • Cerium(III) chloride heptahydrate

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : Cerium chloride heptahydrate

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only