missing translation for 'onlineSavingsMsg'
Learn More
Cerium ingot, 99% (REO), Thermo Scientific Chemicals Product Code.: 11305038

Cerium ingot, 99% (REO), Thermo Scientific Chemicals

Product Code. 11305038
1 kg
Click to view available options
Quantity:
250 g
1 kg
This item is not returnable. View return policy

Product Code. 11305038

Brand: Thermo Scientific Alfa Aesar 043977.A1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under mineral oil
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-45-1
Molecular Formula Ce
Molecular Weight (g/mol) 140.12
MDL Number MFCD00010924
InChI Key GWXLDORMOJMVQZ-UHFFFAOYSA-N
Synonym cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium
PubChem CID 23974
ChEBI CHEBI:33369
IUPAC Name cerium
SMILES [Ce]

Specifications

Color Gray
UN Number UN1333
Quantity 1 kg
Percent Purity 99%
Odor Odorless
Assay (REO)
Chemical Name or Material Cerium ingot

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberT19I008 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog Number043977.A1,043977.30
Lot NumberQ26I037 Certificate TypeCertificate of Analysis Date02/05/2022 Catalog Number043977.A1,043977.30
Lot NumberZ08H103 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog Number043977.A1,043977.30
Lot NumberQ03G041 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number043977.A1,043977.30
Lot NumberL20R027 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog Number043977.A1,043977.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cerium ingot, 99% (REO), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.