Learn More
Cerium rod, 6.35mm (0.25in) dia, 99.9% (metals basis excluding Ta), Thermo Scientific Chemicals
£99.60 - £513.00
Chemical Identifiers
CAS | 7440-45-1 |
---|---|
Molecular Formula | Ce |
Molecular Weight (g/mol) | 140.12 |
MDL Number | MFCD00010924 |
InChI Key | GWXLDORMOJMVQZ-UHFFFAOYSA-N |
Synonym | cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium |
PubChem CID | 23974 |
ChEBI | CHEBI:33369 |
IUPAC Name | cerium |
SMILES | [Ce] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11335108
|
Thermo Scientific Alfa Aesar
044037.HF |
10 mm |
£99.60
10mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11325108
|
Thermo Scientific Alfa Aesar
044037.DJ |
50 mm |
£318.00
50mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11470650
|
Thermo Scientific Alfa Aesar
044037.DM |
100 mm |
£513.00
100mm |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged in mylar under argon
Chemical Identifiers
7440-45-1 | |
140.12 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
23974 | |
cerium |
Ce | |
MFCD00010924 | |
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium | |
CHEBI:33369 | |
[Ce] |
Specifications
Silver | |
Ce | |
UN1333 | |
GWXLDORMOJMVQZ-UHFFFAOYSA-N | |
cerium | |
23974 | |
99.9% | |
Mylar under Argon | |
Cast Rod | |
Cerium rod, ≈6.35mm (0.25 in.) dia. |
7440-45-1 | |
MFCD00010924 | |
cerio, ingot, cerium, chips, foil, 3n, zer, ceramics-aeium, acmc-1bl5r, chips, 3n reo, powder,-200 mesh, cerium | |
[Ce] | |
140.12 | |
CHEBI:33369 | |
Odorless | |
≈0.21g/mm | |
(metals basis excluding Ta) |
Safety and Handling
GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.
P210-P240-P241-P280-P370+P378q
H228
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: CERIUM
missing translation for 'einecsNumber' : 231-154-9
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged in mylar under argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only