missing translation for 'onlineSavingsMsg'
Learn More
Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals Product Code.: 10266510

Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals

Product Code. 10266510 Shop All Thermo Scientific Products
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 10266510

Brand: Thermo Scientific Acros 219190050

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-46-2
Molecular Formula Cs
Molecular Weight (g/mol) 132.91
MDL Number MFCD00134037
InChI Key TVFDJXOCXUVLDH-UHFFFAOYSA-N
Synonym caesium, cesium-133, cesium, elemental, unii-1ksv9v4y4i, metals basis, 1ksv9v4y4i, cesio, zaesium, casium, zasium
PubChem CID 5354618
ChEBI CHEBI:30514
SMILES [Cs]

Specifications

Color Golden
Melting Point 28.5°C
Boiling Point 705.0°C
Density 1.8730g/mL
Assay Percent Range 99.95+%
Quantity 5 g
Merck Index 15, 2007
IUPAC Name caesium
Formula Weight 132.91
Percent Purity ≥99.95%
Grade Trace Metal Basis
Chemical Name or Material Cesium, 99.95+%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0457647 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number219190010,219190050
Lot NumberA0459885 Certificate TypeCertificate of Analysis Date06/02/2024 Catalog Number219190010,219190050
Lot NumberA0457647 Certificate TypeCertificate of Analysis Date17/11/2023 Catalog Number219190010,219190050
Lot NumberA0455537 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number219190010,219190050
Lot NumberA0454889 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number219190010,219190050
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cesium
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
  • Substance which\, in contact with water\, emits flammable gases Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361-Suspected of damaging fertility or the unborn child.
  • H260-In contact with water releases flammable gases which may ignite spontaneously.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P223-Do not allow contact with water.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium, 99.95+%, (trace metal basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.