Learn More
Cesium chloride, 99+%, for analysis, Thermo Scientific Chemicals
£78.19 - £724.61
Chemical Identifiers
CAS | 7647-17-8 |
---|---|
Molecular Formula | ClCs |
Molecular Weight (g/mol) | 168.36 |
MDL Number | MFCD00010955 |
InChI Key | AIYUHDOJVYHVIT-UHFFFAOYSA-M |
Synonym | cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chlorideShow More |
PubChem CID | 24293 |
ChEBI | CHEBI:63039 |
SMILES | [Cl-].[Cs+] |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10531831
|
Thermo Scientific Acros
189500500 |
50 g | Plastic bottle |
£78.19
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10532021
|
Thermo Scientific Acros
189502500 |
250 g | Plastic bottle |
£255.27
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10183820
|
Thermo Scientific Acros
189500010 |
1 kg | Plastic bottle |
£724.61
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7647-17-8 | |
168.36 | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
24293 | |
[Cl-].[Cs+] |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chlorideShow More | |
CHEBI:63039 |
Specifications
1290.0°C | |
7647-17-8 | |
Crystalline Powder | |
ClCs | |
MFCD00010955 | |
cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chloride | |
AIYUHDOJVYHVIT-UHFFFAOYSA-M | |
cesium;chloride | |
24293 | |
168.36 | |
Analytical | |
Cesium chloride, p.a. |
646.0°C | |
White | |
99% min. (Argentometry) | |
CsCl | |
15, 2010 | |
Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone | |
[Cl-].[Cs+] | |
168.36 | |
CHEBI:63039 | |
≥99% | |
3.983 |
Safety and Handling
GHS H Statement
Suspected of damaging fertility or the unborn child.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-600-2
missing translation for 'rtecsNumber' : FK9625000
missing translation for 'tsca' : TSCA
- Cesium chloride
Signal Word
- Warning
Hazard Category
- Reproductive toxicity Category 2
Hazard Statement
- H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.