missing translation for 'onlineSavingsMsg'
Learn More
Cesium chloride, 99+%, for analysis, Thermo Scientific Chemicals Product Code.: 10532021

Cesium chloride, 99+%, for analysis, Thermo Scientific Chemicals

Product Code. 10532021
250 g, Plastic bottle
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10532021

Brand: Thermo Scientific Acros 189502500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-17-8
Molecular Formula ClCs
Molecular Weight (g/mol) 168.36
MDL Number MFCD00010955
InChI Key AIYUHDOJVYHVIT-UHFFFAOYSA-M
Synonym cesium chloride, caesium chloride, cesium monochloride, cesium chloride cscl, dicesium dichloride, cscl, tricesium trichloride, unii-gnr9hml8ba, gnr9hml8ba, caesium 1+ ion chlorideShow MoreShow Less
PubChem CID 24293
ChEBI CHEBI:63039
SMILES [Cl-].[Cs+]

Specifications

Boiling Point 1290.0°C
Melting Point 646.0°C
Color White
Physical Form Crystalline Powder
Quantity 250 g
Assay Percent Range 99% min. (Argentometry)
Linear Formula CsCl
Merck Index 15, 2010
Solubility Information Solubility in water: 1860g/L (20°C). Other solubilities: soluble in alcohol, insoluble in acetone
IUPAC Name caesium(1+) chloride
Formula Weight 168.36
Percent Purity ≥99%
Grade Analytical
Packaging Plastic bottle
Density 3.983
Chemical Name or Material Cesium chloride, p.a.
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466641 Certificate TypeCertificate of Analysis Date17/10/2024 Catalog Number189500010,189500500,189502500
Lot NumberA0462512 Certificate TypeCertificate of Analysis Date07/05/2024 Catalog Number189500010,189500500,189502500
Lot NumberA0449473 Certificate TypeCertificate of Analysis Date18/01/2024 Catalog Number189500010,189500500,189502500
Lot NumberA0456745 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog Number189500010,189500500,189502500
Lot NumberA0455818 Certificate TypeCertificate of Analysis Date25/08/2023 Catalog Number189500010,189500500,189502500
compliance-icons
Product Identifier
  • Cesium chloride
Signal Word
  • Warning
Hazard Category
  • Reproductive toxicity Category 2
Hazard Statement
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cesium chloride, 99+%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.