missing translation for 'onlineSavingsMsg'
Learn More
Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals Product Code.: 10703211

Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals

Product Code. 10703211
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10703211

Brand: Thermo Scientific Acros 205982500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-20-0
Molecular Formula H14O11SZn
Molecular Weight (g/mol) 287.54
MDL Number MFCD00149894
InChI Key RZLVQBNCHSJZPX-UHFFFAOYSA-L
Synonym zinc sulfate heptahydrate, zinc sulfate jan, zinc sulfate 1:1 heptahydrate, unii-n57ji2k7wp, zinc vitriol heptahydrate, zinc sulfate heptahydrate 1:1:7, white vitriol heptahydrate, ccris 5563, zinc sulfate znso4 heptahydrate, znso4.7h2oShow MoreShow Less
PubChem CID 62640
ChEBI CHEBI:32312
IUPAC Name zinc;sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Zn++].[O-]S([O-])(=O)=O

Specifications

Melting Point 100.0°C
Color White
pH 4.4 to 6 (5% soln. at 20°C)
Physical Form Crystalline Powder and/or Crystals
Quantity 250 g
Assay Percent Range 99.4% min. (Complexometry)
Linear Formula ZnSO4·7H2O
Merck Index 15, 10359
Solubility Information Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol
Formula Weight 287.53
Percent Purity 99.5%
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Zinc sulfate heptahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465859 Certificate TypeCertificate of Analysis Date06/09/2024 Catalog Number205980050,205980010,205982500
Lot NumberA0457927 Certificate TypeCertificate of Analysis Date07/02/2024 Catalog Number205980050,205980010,205982500
Lot NumberA0450311 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number205980050,205980010,205982500
Lot NumberA0400590 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number205980050,205980010,205982500
Lot NumberA0442082 Certificate TypeCertificate of Analysis Date14/05/2022 Catalog Number205980050,205980010,205982500
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Zinc sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P273-Avoid release to the environment.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.