missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 100.0°C |
Color | White |
pH | 4.4 to 6 (5% soln. at 20°C) |
Physical Form | Crystalline Powder and/or Crystals |
Quantity | 1 kg |
Assay Percent Range | 99.4% min. (Complexometry) |
Linear Formula | ZnSO4·7H2O |
Merck Index | 15, 10359 |
Solubility Information | Solubility in water: 960g/L. Other solubilities: 1g/2.5mL glycerol, insoluble in alcohol |
Formula Weight | 287.53 |
Show More |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0465859 | Certificate of Analysis | 06/09/2024 | 205980050,205980010,205982500 |
A0457927 | Certificate of Analysis | 07/02/2024 | 205980050,205980010,205982500 |
A0450311 | Certificate of Analysis | 02/05/2023 | 205980050,205980010,205982500 |
A0400590 | Certificate of Analysis | 14/12/2022 | 205980050,205980010,205982500 |
A0442082 | Certificate of Analysis | 14/05/2022 | 205980050,205980010,205982500 |
Safety and Handling



Product Identifier
- Zinc sulfate heptahydrate
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- ACUTE AQUATIC Acute 1
- LONG-TERM AQUATIC HAZARD Chronic 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- H410-Very toxic to aquatic life with long lasting effects.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P273-Avoid release to the environment.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zinc sulfate heptahydrate, 99.5%, for analysis, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction