missing translation for 'onlineSavingsMsg'
Learn More
Copper(II) chloride, 99%, extra pure, anhydrous Product Code.: 10563951

Copper(II) chloride, 99%, extra pure, anhydrous

Product Code. 10563951 Shop All Thermo Scientific Chemicals Products
250 g, Plastic bottle, 250g
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Unit Size:
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 10563951

Brand: Thermo Scientific Acros 206532500

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7447-39-4 | Cl2Cu | 134.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7447-39-4
Molecular Formula Cl2Cu
Molecular Weight (g/mol) 134.45
MDL Number MFCD00010972

Specifications

Boiling Point 993.0°C
Melting Point 498.0°C
Color Brown
Physical Form Fine Crystalline Powder
Quantity 250 g
Assay Percent Range 98.5% min. (CuCl2)
Linear Formula CuCl2
UN Number UN 2802
Fieser 01,163; 02,84; 03,66; 04,105; 05,158; 06,139; 07,79; 08,119; 09,123; 10,106; 11,267; 13,152; 14,100
Merck Index 15, 2623
Synonym Cupric Chloride
Solubility Information Solubility in water: 620g/L (20°C). Other solubilities: soluble in alcohol, acetone and ether
Formula Weight 134.45
Percent Purity 99%
Grade Extra Pure
Packaging Plastic bottle
Chemical Name or Material Copper(II) Chloride, Anhydrous
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0474086 Certificate TypeCertificate of Analysis Date29/07/2025 Product Code206532500,206530025,206530010
Lot NumberA0470749 Certificate TypeCertificate of Analysis Date17/05/2025 Product Code206532500,206530025,206530010
Lot NumberA0461834 Certificate TypeCertificate of Analysis Date03/05/2024 Product Code206532500,206530025,206530010
Lot NumberA0437623 Certificate TypeCertificate of Analysis Date16/02/2024 Product Code206532500,206530025,206530010
Lot NumberA0450039 Certificate TypeCertificate of Analysis Date15/02/2024 Product Code206532500,206530025,206530010
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Copper(II) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper(II) chloride, 99%, extra pure, anhydrous >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.