Learn More
Cesium metavanadate, 99.9% (metals basis), Thermo Scientific Chemicals
£51.70 - £234.00
Chemical Identifiers
CAS | 14644-55-4 |
---|---|
Molecular Formula | CsO3V |
Molecular Weight (g/mol) | 231.844 |
MDL Number | MFCD00049812 |
InChI Key | SZIZYHNGMIYCQE-UHFFFAOYSA-N |
Synonym | Cesium vanadate, meta |
PubChem CID | 23689378 |
IUPAC Name | cesium;oxido(dioxo)vanadium |
SMILES | [O-][V](=O)=O.[Cs+] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11313608
|
Thermo Scientific Alfa Aesar
039213.06 |
5 g | |||||||
11323608
|
Thermo Scientific Alfa Aesar
039213.14 |
25 g |
£234.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is employed as a intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
14644-55-4 | |
231.844 | |
SZIZYHNGMIYCQE-UHFFFAOYSA-N | |
23689378 | |
[O-][V](=O)=O.[Cs+] |
CsO3V | |
MFCD00049812 | |
Cesium vanadate, meta | |
cesium;oxido(dioxo)vanadium |
Specifications
14644-55-4 | |
Powder | |
MFCD00049812 | |
Cesium vanadate, meta | |
[O-][V](=O)=O.[Cs+] | |
231.844 | |
231.84 | |
Odorless | |
Cesium metavanadate |
White to Yellow | |
CsO3V | |
UN3288 | |
SZIZYHNGMIYCQE-UHFFFAOYSA-N | |
cesium;oxido(dioxo)vanadium | |
23689378 | |
99.9% | |
(metals basis) |
Safety and Handling
GHS H Statement
H370-H302-H332
Causes damage to organs.
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P260-P261-P304+P340-P307+P311-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF exposed: Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only