missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CHAPS (White Crystalline Powder), Fisher BioReagents™
£69.89 - £108.97
Chemical Identifiers
CAS | 75621-03-3 |
---|---|
Molecular Formula | C32H58N2O7S |
Molecular Weight (g/mol) | 614.883 |
InChI Key | UMCMPZBLKLEWAF-GBSSQDIOSA-N |
Synonym | 3-[(3-Cholamidopropyl) dimethylammonio]-1-propanesulfonate |
PubChem CID | 134129639 |
IUPAC Name | 3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate |
SMILES | CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C |
Description
CHAPS is a Zwitterionic detergent used for solubilizing membrane proteins.Chemical Identifiers
75621-03-3 | |
614.883 | |
3-[(3-Cholamidopropyl) dimethylammonio]-1-propanesulfonate | |
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate |
C32H58N2O7S | |
UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
134129639 | |
CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C |
Specifications
0.1 max. (1%) at 260nm, 0.1 max. (1%) at 280nm | |
75621-03-3 | |
267°C | |
≥98 % | |
15, 2043 | |
UMCMPZBLKLEWAF-GBSSQDIOSA-N | |
3-[dimethyl-[3-[[(4R)-4-[(3R,5S,7R,8R,9R,10S,12S,13R,14R,17S)-3,7,12-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoyl]amino]propyl]azaniumyl]propane-1-sulfonate | |
134129639 | |
CHAPS |
White | |
Solid | |
Pass Test | |
C32H58N2O7S | |
3-[(3-Cholamidopropyl) dimethylammonio]-1-propanesulfonate | |
CC(CCC(=O)NCCC[N+](C)(C)CCCS(=O)(=O)[O-])C1CCC2C1(C(CC3C2C(CC4C3(CCC(C4)O)C)O)O)C | |
614.883 | |
614.9 |
Safety and Handling
WARNING!
Emergency Overview
Irritating to eyes, respiratory system and skin. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Obtain medical attention. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Obtain medical attention. Do not induce vomiting. Obtain medical attention. . .
NFPA
Health:2
Flammability:1
Instability:0
Recommended Storage : RT