missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand: Thermo Scientific™ 201249
This item is not returnable.
View return policy
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Features:- Seven-day recorder
- Functions to -150°C
- Field or factory installed
Ordering information:
- Field-installed units must be installed by a qualified professional
Specifications
Inkless Chart Recorder, Factory Installed | |
-150°C | |
7 Days |
Chart Recorder | |
Forma™ 900, HERAfreeze™ HFU-B, Revco™ ExF/DxF, TSE/TSD Series | |
Recorder |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-5 of 6
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers > Inkless Chart Recorder; Factory installed; Includes box of 50 chart papers; For use with Forma 900 and 7000, HERAfreeze HFU B, Revco ExF/DxF, and TSE/TSD series
Spot an opportunity for improvement?Share a Content Correction