Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand:  Thermo Scientific™ FCRP567TSX
 View more versions of this product
Product Code. 15369615
  • £1712.14 / Each
Estimated Shipment: 19-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Features:
  • Seven-day recorder
  • Field or factory installed
Ordering information:
  • Field-installed units must be installed by a qualified professional
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ink Chart Recorder, Field Installed
-100°C to +38°C
7 Days
Chart Recorder
TSX Series
Recorder