Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand:  Thermo Scientific™ FCR600TSXLE
 View more versions of this product
Product Code. 15728537
  • £1712.14 / Each
Estimated Shipment: 14-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed ink chart recorder
Services
Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inkless Chart Recorder, Field Installed
Chart Recorder
TSX600 ultra-low freezers
Recorder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers >