Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand:  Thermo Scientific™ FCRP500LE
 View more versions of this product
Product Code. 15768537
  • £1712.14 / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed inkless chart recorder
Services
Warranty varies by country, please contact us for details
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ink Chart Recorder, Field Installed
Chart Recorder
Revco RLE, Forma 89000, TLE Series, HLE Series (500 models only)
Recorder
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers >