Learn More
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers
Record seven days of temperature performance at temperatures to -150° C.
Brand: Thermo Scientific™ FCRP600LE
This item is not returnable.
View return policy
Description
Record seven days of temperature performance at temperatures to -150° C using Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers. Recorders use 6 in. (15cm) chart paper and are available as factory or field installed.
Field-installed inkless chart recorderServices
Warranty varies by country, please contact us for details
Specifications
Ink Chart Recorder, Field Installed | |
Revco RLE, Forma 89000, TLE Series, HERAfreeze HLE 600-box freezers | |
Boxes (600) |
Chart Recorder | |
Recorder |
Product Content Correction
Product Title
Thermo Scientific™ Chart Recorders for Ultra-Low Temperature Freezers >
Spot an opportunity for improvement?Share a Content Correction