missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Human CHEK2 (CHK2), His Tag Recombinant Protein

Human CHEK2 (CHK2), His Tag Recombinant Protein

Brand:  Thermo Scientific™ PV3983

Code : 50

Additional Details : Weight : 0.05250kg

 View more versions of this product

Product Code. 10003492

  • £6900.00 / 1mg
Estimated Shipment: 12-08-2024
to see stock.

For Research Use Only. All usage must comply with product instructions.
 

Description

Description

  • CHEK2 (CHK2) is a serine/threonine kinase and a component of the DNA damage checkpoint pathway
  • CHK2 is activated by DNA damage and phosphorylates several modulators of cell cycle control including tumor suppressor proteins
  • A mutation in this gene has been linked to cancer
Specifications

Specifications

NP_009125
66 kDa
CHEK2 (CHK2)
Human
His Tag
Ser/Thr Kinases: CAMK Group
Baculovirus
Frozen
Kinase Assay
1 mg
RAD53 Kinase Family
Recombinant
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.04% Triton X-100, 4mM DTT, 50% glycerol
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Dry Ice
Full Length
Drug Development
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers