missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Human CHEK2 (CHK2), His Tag Recombinant Protein
Human CHEK2 (CHK2), His Tag Recombinant Protein
£6900.00
Specifications
Accession Number | NP_009125 |
---|---|
For Use With (Application) | Kinase Assay |
Molecular Weight (g/mol) | 66 kDa |
Gene Alias | CHEK2 (CHK2) |
KinaseFamily | RAD53 Kinase Family |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
10003492
|
Thermo Scientific™
PV3983 |
1 mg |
£6900.00
1mg |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- CHEK2 (CHK2) is a serine/threonine kinase and a component of the DNA damage checkpoint pathway
- CHK2 is activated by DNA damage and phosphorylates several modulators of cell cycle control including tumor suppressor proteins
- A mutation in this gene has been linked to cancer
Specifications
NP_009125 | |
66 kDa | |
RAD53 Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.04% Triton X-100, 4mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Baculovirus |
Kinase Assay | |
CHEK2 (CHK2) | |
Human | |
His Tag | |
Dry Ice |