Learn More
CHEMetrics CHEMets Dissolved Oxygen, Visual Water Testing Kit
Provides results in two minutes or less
Brand:  CHEMetrics K-7540
 View more versions of this product
Product Code. 11731004
  • £62.60 / Pack of 30
Explore more special offers
Includes: REMOVE
Description
Description
Provide results in two minutes or less
  • Designed for maximum simplicity and accuracy, Chemets 7 mm diameter ampules feature tapered, prescored tips with reagent vacuum-sealed inside
  • To perform a test, simply immerse Chemets ampule in the sample, snap off tip and the correct sample volume is quickly drawn inside the ampule
  • In two minutes or less, the reaction between the sample and reagent yields a color—which can then be compared to appropriate color standard for quantification of results
  • Comparators have two-year shelf life.
Specifications
Specifications
Dissolved Oxygen (DO)
30
2 min.
Plastic case, Refill, Comparator, adhesive mounting clamp, permanent mounting clamp, sampling tube and instructions
0 to 40 ppb
CHEMets Visual Water Testing Kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS Documents
Documents
Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
CHEMetrics CHEMets Dissolved Oxygen, Visual Water Testing Kit > Dissolved Oxygen: Rhodazine D method; 0 to 40ppb