Learn More
Medimark International™ Chemgene™ HLD4L Laboratory Disinfectant
For multipurpose surface disinfection and cleaning. Chemgene™ HLD4L Lab Disinfectant destroys bacteria, fungi, mycobacteria, viruses and spores.
Brand: Medimark International™ XTM309
Code : 04
Additional Details : CAS Number : 51981-21-6 Weight : 4.95000kg Transport : UN number : 1760 Chem class : 8 Pack group : III
Description
For multipurpose surface disinfection and cleaning. Chemgene™ Lab Disinfectant Spray destroys bacteria, fungi, mycobacteria, viruses and spores.
Chemgene HLD4L disinfectant concentrate. 5L container.- Manufactured in accordance with Quality Management System ISO 9001
- Biodegradable under OECD conditions
- Denatures DNA/RNA
Specifications
Lab Disinfectant | |
ISO 9001 | |
For multipurpose Surface disinfection and cleaning |
Clear | |
0.048611111111 | |
5 L |
Safety and Handling
- Chemgene HLD4L disinfectant concentrate
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P102-Keep out of reach of children.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P315-Get immediate medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : didecyldimethylammonium chloride, quaternary ammonium compounds, benzyl-C8-18- alkyldimethyl, chlori