Learn More
Medimark International™ Chemgene™ HLD4L Laboratory Disinfectant
For multipurpose surface disinfection and cleaning. Chemgene™ HLD4L Lab Disinfectant destroys bacteria, fungi, mycobacteria, viruses and spores.
£7.50 - £49.60
Specifications
Appearance Clear
View More Specs
Products 3
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
15106686
View Documents
Medimark International™
XTM301
750 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15136686
View Documents
Medimark International™
XTM305
1 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15126686
View Documents
Medimark International™
XTM309
5 L This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
For multipurpose surface disinfection and cleaning. Chemgene™ Lab Disinfectant Spray destroys bacteria, fungi, mycobacteria, viruses and spores.
Chemgene HLD4L disinfectant concentrate.
  • Cleans and disinfects all areas of the laboratory
  • Manufactured in accordance with Quality Management System ISO 9001
  • Biodegradable under OECD conditions
  • Denatures DNA/RNA
  • Can be used at a dilution of: 1 in 100, General conditions; or 1 in 10 for Bio-hazard contamination
Specifications
Specifications
Clear
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Medimark International™ Chemgene™ HLD4L Laboratory Disinfectant