Learn More
Sampling Systems™ Chemical Spoon and Spatula
This versatile chemical spoon and spatula has a robust design and is made from 304 stainless steel. It can be used as both a spoon and a spatula to allow for greater versatility in the sampling of both powder and liquid products.
Brand: Sampling Systems™ A625-280
Additional Details : Weight : 0.11000kg
Description
- No sharp edges or crevices, making it ideal for use in food, pharma and chemical companies
- Available in a range of sizes
- Heavy duty design for comfortable use
- Ideal for measuring, sampling and dosing powders and liquids
Specifications
Spoon | |
Autoclavable | |
No | |
280 mm |
Stainless Steel | |
Reusable | |
Non-sterile |