Learn More
Liebherr™ Chest Freezer
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ Chest Freezers feature aluminum interior containers and stainless steel adjustable feet.
Brand:  Liebherr™ GTL 6105
 View more versions of this product
Product Code. 12014672
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Steel chest freezer
  • Mechanic control system
  • External Analog display
  • Ergonomic handle with aluminum trim
  • Fitted lock
Specifications
Specifications
601 L
White
91.7 H x 164.7 W x 77.6 D cm
70.2 cm
220/240 V
R600A
-14°C to -26°C
GTL6105
57.5 cm
152 cm
50 Hz