Learn More
Chlorobenzene-d{5}, 99% (Isotopic), Thermo Scientific Chemicals
£70.12 - £282.41
Chemical Identifiers
CAS | 3114-55-4 |
---|---|
Molecular Formula | C6H5Cl |
Molecular Weight (g/mol) | 117.587 |
MDL Number | MFCD00000531 |
InChI Key | MVPPADPHJFYWMZ-RALIUCGRSA-N |
Synonym | chlorobenzene-d5, chloro 2h5 benzene, 1-chloro 2 h? benzene, chlorobenzene d5, benzene-d5, chloro, 2h5 phenyl chloride, 1-chloro 2h5 benzene, benzene-d5-, chloro, chlorobenzene, perdeutero, chlorobenzene-d5, 99 atom % d |
PubChem CID | 575975 |
IUPAC Name | 1-chloro-2,3,4,5,6-pentadeuteriobenzene |
SMILES | C1=CC=C(C=C1)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11439940
|
Thermo Scientific Alfa Aesar
044712.03 |
1 g | N/A | ||||||
11449940
|
Thermo Scientific Alfa Aesar
044712.06 |
5 g |
£282.41
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlorobenzene-d5 is a halogenated D5 chlorobenzene for proteomics research. It is a precursor to deuterated dyes and rubbers. It is used as a high-boiling solvent in industrial applications.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
3114-55-4 | |
117.587 | |
MVPPADPHJFYWMZ-RALIUCGRSA-N | |
575975 | |
C1=CC=C(C=C1)Cl |
C6H5Cl | |
MFCD00000531 | |
chlorobenzene-d5, chloro 2h5 benzene, 1-chloro 2 h? benzene, chlorobenzene d5, benzene-d5, chloro, 2h5 phenyl chloride, 1-chloro 2h5 benzene, benzene-d5-, chloro, chlorobenzene, perdeutero, chlorobenzene-d5, 99 atom % d | |
1-chloro-2,3,4,5,6-pentadeuteriobenzene |
Specifications
Liquid | |
24°C (75°F) | |
1.522 | |
130°C | |
MFCD00000531 | |
1866335 | |
MVPPADPHJFYWMZ-RALIUCGRSA-N | |
1-chloro-2,3,4,5,6-pentadeuteriobenzene | |
575975 | |
99% |
1.157g/mL | |
Aromatic | |
3114-55-4 | |
C6H5Cl | |
UN1134 | |
chlorobenzene-d5, chloro 2h5 benzene, 1-chloro 2 h? benzene, chlorobenzene d5, benzene-d5, chloro, 2h5 phenyl chloride, 1-chloro 2h5 benzene, benzene-d5-, chloro, chlorobenzene, perdeutero, chlorobenzene-d5, 99 atom % d | |
C1=CC=C(C=C1)Cl | |
117.587 | |
117.6 | |
Chlorobenzene-d5, Isotopic |
Safety and Handling
GHS H Statement
H226-H332
Flammable liquid and vapor.
Harmful if inhaled.
GHS P Statement
P262-P273
Do not get in eyes, on skin, or on clothing.
Avoid release to the environment.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
RUO – Research Use Only