missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chloroform, ≥99.5%, Honeywell™
Contains 100-200 ppm amylenes as stabilizer,≥99.5%
£34.37 - £349.70
Chemical Identifiers
CAS | 67-66-3 |
---|---|
Molecular Formula | CHCl3 |
Molecular Weight (g/mol) | 119.37 |
MDL Number | MFCD00000826 |
InChI Key | HEDRZPFGACZZDS-UHFFFAOYSA-N |
Synonym | trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio |
PubChem CID | 6212 |
ChEBI | CHEBI:35255 |
SMILES | ClC(Cl)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
15668480
|
Honeywell Research Chemicals
C2432-25ML |
25 mL | Glass bottle |
£34.37
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15677730
|
Honeywell Research Chemicals
C2432-4X25ML |
4 x 25 mL | Glass bottle |
£128.17
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15687730
|
Honeywell Research Chemicals
C2432-500ML |
500 mL | Glass bottle |
£75.75
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15657730
|
Honeywell Research Chemicals
C2432-1L |
1 L | Glass bottle |
£91.36
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15667730
|
Honeywell Research Chemicals
C2432-2.5L |
2.5 L | Glass bottle |
£163.59
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15697730
|
Honeywell Research Chemicals
C2432-6X500ML |
6 x 500 mL | Glass bottle |
£349.70
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Chemical Identifiers
67-66-3 | |
119.37 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
6212 | |
ClC(Cl)Cl |
CHCl3 | |
MFCD00000826 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
CHEBI:35255 |
Specifications
67-66-3 | |
Colorless | |
61°C (1013 hPa) | |
CHCl3 | |
UN1888 | |
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio | |
ClC(Cl)Cl | |
Amylenes | |
6212 | |
CHEBI:35255 | |
160mmHg (20°C) | |
Liquid |
-61.3°C | |
1.492g/mL at 25°C | |
Sweet | |
MFCD00000826 | |
1731042 | |
HEDRZPFGACZZDS-UHFFFAOYSA-N | |
chloroform | |
119.37 | |
4.1 (vs air) | |
119.38g/mol | |
≥99.5% | |
Chloroform |
Safety and Handling
P261-P281-P305 + P351 + P338-P311
missing translation for 'dotInformation' : 6.1/PGIII
missing translation for 'rtecsNumber' : 200-663-8
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Chloroform
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Acute toxicity Category 3
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- Specific target organ toxicity Category 1
- Carcinogenicity Category 2
- Reproductive toxicity Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H361d-Suspected of damaging the unborn child.
- H372-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.