missing translation for 'onlineSavingsMsg'
Learn More
Chloroform, Reagent ACS, Spectro Grade, 99.8%, Thermo Scientific Chemicals Product Code.: 10547552

Chloroform, Reagent ACS, Spectro Grade, 99.8%, Thermo Scientific Chemicals

Product Code. 10547552
500 mL, EcoSafPak™ Glass Bottle
Click to view available options
Quantity:
500 mL
2.5 L
Packaging:
EcoSafPak™ Glass Bottle
Glass Bottle
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 10547552

Brand: Thermo Scientific Acros 404635000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XVII to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For UV/VIS spectroscopy
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-66-3, 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformioShow MoreShow Less
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl

Specifications

Melting Point -63.0°C
Absorbance 0.01 max. at 290nm, 0.01 max. at 400nm, 0.05 max. at 270nm, 0.15 max. at 260nm, 0.25 max. at 255nm, 1 max. at 245nm
Decomposition Information 290°C
Density 1.4800g/mL
Boiling Point 61.0°C
Quantity 500 mL
Packaging EcoSafPak™ Glass Bottle
Physical Form Liquid
Refractive Index 1.4443 to 1.4463
Assay Percent Range 99.8%
Residue after Evaporation 0.001% max.
Specific Gravity 1.48
Linear Formula CHCl3
Viscosity 0.56 mPa.s (20°C)
Beilstein 01, 61
Fieser 01,130; 08,92; 15,84
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
IUPAC Name chloroform
Formula Weight 119.38
Percent Purity 99.8%
Grade Spectroscopy
Chemical Name or Material Chloroform
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467877 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number404635000,404630025
Lot NumberA0450347 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number404635000,404630025
Lot NumberB0541096B Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number404635000,404630025
Lot NumberB0542007 Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number404635000,404630025
Lot NumberB0542684A Certificate TypeCertificate of Analysis Date14/11/2023 Catalog Number404635000,404630025
compliance-icons compliance-icons
Product Identifier
  • Chloroform, stabilized with ethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H372-Causes damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P311-Call a POISON CENTER/doctor/ .
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contains : Chloroform

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroform, Reagent ACS, Spectro Grade, 99.8%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.