missing translation for 'onlineSavingsMsg'
Learn More

Chloroform, stabilized with ethanol, for HPLC, Thermo Scientific Chemicals

Chloroform, >-99%, CHCl3, CAS Number-67-66-3, 64-17-5, trichloromethane, trichloroform, trichlormethan, methyl trichloride, cloroformio, methenyl trichloride, formyl trichloride, methane, trichloro, chloroforme, methane trichloride, 2.5L

£37.39 - £68.24

Chemical Identifiers

CAS 67-66-3, 64-17-5
Molecular Formula CHCl3
Molecular Weight (g/mol) 119.37
MDL Number MFCD00000826
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
Synonym trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
PubChem CID 6212
ChEBI CHEBI:35255
SMILES ClC(Cl)Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10289473
View Documents Promotion Details
Thermo Scientific Acros
390760010
1 L Glass Bottle
£37.39
1 L
Please to purchase this item. Need a web account? Register with us today!
11360856
View Documents
Thermo Scientific Acros
390760025
2.5 L Glass Bottle
£68.24
2.50 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For HPLC
Specifications

Chemical Identifiers

67-66-3, 64-17-5
119.37
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
ClC(Cl)Cl
CHCl3
MFCD00000826
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
CHEBI:35255

Specifications

-63.0°C
67-66-3, 64-17-5
1.4800g/mL
Crystals
99+%
1.48
CHCl3
0.56 mPa.s (20°C)
15, 2142
Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in aceton
ClC(Cl)Cl
119.37
CHEBI:35255
≥99%
Chloroform, Stabilized with ethanol
0.009 max. at 300nm, 0.046 max. at 265nm, 0.301 max. at 250nm, 0.699 max. at 245nm, 1 cm cell vs water, 1 cm cell vs water at 0nm
White
61.0°C
1.4443 to 1.4463
0.0002 % max.
CHCl3
MFCD00000826
0.01% Max. (K.F.)
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane
6212
119.38
HPLC
Videos
Safety and Handling

Safety and Handling

GHS P Statement Harmful if swallowed. Toxic if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

Product Identifier
  • Chloroform, stabilized with ethanol

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Carcinogenicity Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H302-Harmful if swallowed.
  • H331-Toxic if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H361d-Suspected of damaging the unborn child.
  • H336-May cause drowsiness or dizziness.
  • H372-Causes damage to organs.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : Chloroform, Ethyl alcohol

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.