Learn More
Chlorogenic acid, Thermo Scientific Chemicals
An analog of caffeic acid
£30.15 - £244.22
Chemical Identifiers
CAS | 327-97-9 |
---|---|
Molecular Formula | C16H18O9 |
Molecular Weight (g/mol) | 354.31 |
MDL Number | MFCD00003862 |
InChI Key | CWVRJTMFETXNAD-JUHZACGLSA-N |
Synonym | chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri |
PubChem CID | 1794427 |
ChEBI | CHEBI:16112 |
IUPAC Name | (1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid |
SMILES | O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15443229
|
Thermo Scientific Alfa Aesar
J60457.MD |
250 mg |
£30.15
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15423229
|
Thermo Scientific Alfa Aesar
J60457.03 |
1 g |
£67.54
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15433229
|
Thermo Scientific Alfa Aesar
J60457.06 |
5 g |
£244.22
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorogenic acid is used as a food additive in coffee products, chewing gum and mint. It acts as an intermediate in the biosynthesis of lignin, which is an antioxidant.
Solubility
Soluble in ethanol and acetone.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
327-97-9 | |
354.31 | |
CWVRJTMFETXNAD-JUHZACGLSA-N | |
1794427 | |
(1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid |
C16H18O9 | |
MFCD00003862 | |
chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri | |
CHEBI:16112 | |
O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O |
Specifications
327-97-9 | |
C16H18O9 | |
14,2142 | |
Soluble in ethanol and acetone. | |
O[C@@H]1C[C@](O)(C[C@@H](OC(=O)\C=C\C2=CC=C(O)C(O)=C2)[C@@H]1O)C(O)=O | |
354.31 | |
CHEBI:16112 | |
Powder |
210°C (decomposition) | |
MFCD00003862 | |
chlorogenic acid, 3-caffeoylquinic acid, chlorogenate, 3-o-caffeoylquinic acid, 3-3,4-dihydroxycinnamoyl quinic acid, heriguard, hlorogenic acid, caffeoyl quinic acid, 3-caffeoylquinate, unii-318adp12ri | |
CWVRJTMFETXNAD-JUHZACGLSA-N | |
(1S,3R,4R,5R)-3-[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy-1,4,5-trihydroxycyclohexane-1-carboxylic acid | |
1794427 | |
354.31 | |
Chlorogenic acid |
Safety and Handling
missing translation for 'einecsNumber' : 206-325-6
missing translation for 'rtecsNumber' : GU8480000
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only