missing translation for 'onlineSavingsMsg'
Learn More
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals Product Code.: 11400244

Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals

Product Code. 11400244
100 mL
Click to view available options
Quantity:
25 mL
100 mL
500 mL
This item is not returnable. View return policy

Product Code. 11400244

Brand: Thermo Scientific Alfa Aesar A13651.AE

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chlorotrimethylsilane is used to silanize laboratory glassware, which makes the surfaces more lipophilic. It is used in the production of trimethylsilyl halides, pseudohalides and various organic silicon compounds. It is also used to produce hexamethyldisilane by reduction.

Solubility
Miscible with ether, benzene, diethylether and perchloroethylene.

Notes
Incompatible with water, strong acids, strong oxidizing agents, strong bases, alcohols, aldehydes, amines, ketones and esters.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-77-4
Molecular Formula C3H9ClSi
Molecular Weight (g/mol) 108.64
MDL Number MFCD00000502
InChI Key IJOOHPMOJXWVHK-UHFFFAOYSA-N
Synonym trimethylchlorosilane, trimethylsilyl chloride, silane, chlorotrimethyl, trimethyl chlorosilane, monochlorotrimethylsilicon, tmcs, chloro trimethyl silane, silane, trimethylchloro, silicane, chlorotrimethyl, tmsclShow MoreShow Less
PubChem CID 6397
ChEBI CHEBI:85069
IUPAC Name chloro(trimethyl)silane
SMILES C[Si](C)(C)Cl

Specifications

Boiling Point 56°C to 58°C
Melting Point -58°C
Quantity 100 mL
Linear Formula (CH3)3SiCl
UN Number UN1298
Beilstein 1209232
Flash Point −27°C (−16°F)
Solubility Information Miscible with ether,benzene,diethylether and perchloroethylene.
Formula Weight 108.64
Percent Purity ≥98%
Odor Chlorine-like
Refractive Index 1.389
Sensitivity Moisture sensitive
Density 0.856
Chemical Name or Material Chlorotrimethylsilane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462601 Certificate TypeCertificate of Analysis Date02/12/2024 Catalog NumberA13651.AP,A13651.AE,A13651.AU,A13651.AC
Lot NumberA0459555 Certificate TypeCertificate of Analysis Date13/03/2024 Catalog NumberA13651.AP,A13651.AE,A13651.AU,A13651.AC
Lot Number10247424 Certificate TypeCertificate of Analysis Date19/07/2023 Catalog NumberA13651.AP,A13651,A13651.AE,A13651.AU,A13651.AC
Lot Number10245026 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog NumberA13651.AP,A13651.AE,A13651.AU,A13651.AC
Lot NumberA0446436 Certificate TypeCertificate of Analysis Date24/11/2022 Catalog NumberA13651.AP,A13651.AE,A13651.AU,A13651.AC
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Chlorotrimethylsilane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • EUH014-Reacts violently with water.
  • EUH071-Corrosive to the respiratory tract.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Supplemental information
  • MIXTURE LIST-Contains : Trimethylchlorosilane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chlorotrimethylsilane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.