Learn More
Chlorotriphenylsilane, 96%, Thermo Scientific Chemicals
£21.40 - £199.00
Chemical Identifiers
CAS | 76-86-8 |
---|---|
Molecular Formula | C18H15ClSi |
Molecular Weight (g/mol) | 294.85 |
MDL Number | MFCD00000496 |
InChI Key | MNKYQPOFRKPUAE-UHFFFAOYSA-N |
Synonym | triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl |
PubChem CID | 6458 |
IUPAC Name | chloro(triphenyl)silane |
SMILES | Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11420294
|
Thermo Scientific Alfa Aesar
A13678.06 |
5 g |
£21.40
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11430294
|
Thermo Scientific Alfa Aesar
A13678.14 |
25 g |
£59.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11423900
|
Thermo Scientific Alfa Aesar
A13678.18 |
50 g |
£97.10
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11440294
|
Thermo Scientific Alfa Aesar
A13678.22 |
100 g |
£199.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsChlorotriphenylsilane is used as a silylating agent. It is also used as a pharmaceutical intermediate.
Solubility
Reacts with water.
Notes
Keep container tightly sealed. Store under dry inert gas. It is sensitive to moisture. Incompatible with bases and oxidizing agents.
Chemical Identifiers
76-86-8 | |
294.85 | |
MNKYQPOFRKPUAE-UHFFFAOYSA-N | |
6458 | |
Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 |
C18H15ClSi | |
MFCD00000496 | |
triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl | |
chloro(triphenyl)silane |
Specifications
188°C to 189°C (3mmHg) | |
76-86-8 | |
MFCD00000496 | |
1820487 | |
triphenylsilyl chloride, triphenylchlorosilane, silane, chlorotriphenyl, triphenylsilicon chloride, chloro triphenyl silane, triphenylsilylchloride, chloro-tri phenyl silane, benzene, 1,1',1-chlorosilylidyne tris, clsiph3, ph3sicl | |
MNKYQPOFRKPUAE-UHFFFAOYSA-N | |
chloro(triphenyl)silane | |
6458 | |
96% | |
Moisture sensitive |
91°C to 96°C | |
C18H15ClSi | |
UN3261 | |
>200°C (392°F) | |
Reacts with water. | |
Cl[Si](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1 | |
294.85 | |
294.86 | |
Pungent | |
Chlorotriphenylsilane |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 200-989-0
missing translation for 'rtecsNumber' : VV2720000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only