missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ CHO CD EfficientFeed™ B AGT™ Nutrient Supplement

CHO CD EfficientFeed™ B AGT™ Nutrient Supplement

Brand:  Gibco™ A1245601

Code : 50

Additional Details : Weight : 113.39000kg

 View more versions of this product

Product Code. 11370892

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Gibco™ CHO CD EfficientFeed™ B AGT™ nutrient supplement is a single part dry format product for fed batch addition to the culture of multiple cell lines. CHO CD EfficientFeed™ B AGT™ (Advanced Granulation Technology™) nutrient supplement is an animal origin-free, chemically defined formulation that contains no proteins, hydrolysates, or components of incompletely defined composition.

The AGT™ format is designed for large scale manufacturing and ease of use. This product is also available in liquid format for small scale use and experimentation.
  • Expect up to 2- to 5-fold increases in productivity
  • Avoid issues associated with the use of hydrolysates and other alternatives while gaining superior performance
  • Simplify scale-up by leveraging the value of AGT™ simple reconstitution
  • Achieve performance comparable to our original liquid catalog product
  • Ready for larger scale use with convenient packaging options (10 L and 100 L)

    Which EfficientFeed™ Nutrient Supplement to Choose?
    Consider the CHO CD Efficient Feed™ Kit when:
  • You are using a base medium with less than 6 g/L glucose and no hydrolysate at day 0

    Consider CHO CD EfficientFeed™ A supplement when:
  • You are currently using CD OptiCHO™ as your base cell growth medium

    Consider CHO CD EfficientFeed™ B supplement when:
  • You are currently using CD CHO as your base cell growth medium

    Consider CD EfficientFeed™ C AGT™ supplement when:
  • You want higher productivity in GS CHO and other selected cell lines
  • You are currently using CD FortiCHO™ as your base cell growth medium
  • You are using a base medium with greater than 6 g/L glucose or a hydrolysate at day 0

    Increased Titers
    CHO CD EfficientFeed™ supplements have been shown to increase titers 2-5 fold versus batch culture, and are an excellent starting point for process development (Figures 1a & 1b). Furthermore, they outperform competitive alternatives, including hydrolysates (Figures 2, 3a, & 3b).

    Simple Scale-Up and Cost Savings
    The ability to scale up in a dry format provides ease of handling and storage in manufacturing and other bioprocess settings. With AGT™ (Advanced Granulation Technology™) dry format, it is possible to store and use the product even more easily and safely than traditional dry format products. CHO CD EfficientFeed™ B AGT™ nutrient supplement is offered in 10 L and 100 L packaging that allows scale-up from the 1 L catalog liquid as a seamless process, as well as allow for cost savings when purchasing production level volumes.

    Comparable Performance to Liquid Supplement
    Equally important as the convenience and simplicity of AGT™ (Advanced Granulation Technology™) dry format is the comparable performance to the liquid CHO CD EfficientFeed™ B (Figure 4). Both forms have performed similarly in our testing, and the AGT™ version has been used successfully as a prototype in many processes by multiple companies.

    AGT™ Format Ease of Use
    CHO CD EfficientFeed™ B AGT™ nutrient supplement delivers dry format scale-up convenience as well as simple reconstitution via water addition and a single pH adjustment step (see Product Profile Sheet below for complete instructions).

    For research or further manufacturing use. Not for use in diagnostic procedures.
  • Specifications

    Specifications

    Dry format medium
    Store at 2 to 8°C, Store Dark and Dry
    Animal Origin-free, Chemically-defined, Protein-free, Serum-free
    AGT
    Serum-Free
    Sodium Pyruvate, Sodium Bicarbonate
    Low
    Cell Culture Grade, Qualified for Further Manufacturing, Research Grade
    Chemically Defined Nutrient Feeding Supplement
    Solubility
    CHO
    Suspension Cell Culture
    Nutrient Supplement
    Non-sterile
    No Glutamine, No Phenol Red, No Insulin
    Ropak Bucket
    100 L
    Wet Ice
    Product Suggestions

    Product Suggestions

    Videos
    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers

    For Research Use or Further Manufacturing. Not for use in diagnostic procedures.