Learn More
Thermo Scientific Chemicals Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized
Brand: Thermo Scientific Chemicals 386721000
30.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.
Code : 29
Additional Details : CAS Number : 123-41-1 Weight : 0.10700kg Transport : UN number : 3267 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Choline hydroxide, 45 wt.% aqueous solution | |
White | |
Glass bottle | |
MFCD00002831 | |
1.07 | |
choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide | |
KIZQNNOULOCVDM-UHFFFAOYSA-M | |
2-hydroxyethyl(trimethyl)azanium;hydroxide | |
1800ppm ethylenediamine 3000ppm hydroxylamine | |
121.18 | |
Pure |
123-41-1,7732-18-5 | |
1.0700g/mL | |
C5H15NO2 | |
100 mL | |
15,2211 | |
Solubility in water: miscible. | |
[OH-].C[N+](C)(C)CCO | |
121.18 | |
31255 | |
44.0 to 48.0% (Total base) | |
Granular Powder |
Safety and Handling
- Choline hydroxide, stabilized, 45wt.% aqueous solution
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Choline hydroxide
RUO – Research Use Only