missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized
£37.40 - £1580.00
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10128512
|
Thermo Scientific Chemicals
386721000 |
100 mL | Glass bottle |
£37.40
100mL |
Estimated Shipment: 12-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
10582683
|
Thermo Scientific Chemicals
386725000 |
500 mL | Glass Bottle |
£85.20
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Currently Unavailable: We have no information when this product will be available
View alternative products
|
11386725
|
Thermo Scientific Chemicals
386720025 |
2.5 L | Glass bottle |
£293.00
2.50 L |
Estimated Shipment: 22-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
11945441
|
Thermo Scientific Chemicals
386720100 |
10 L | Plastic drum |
£1580.00
10 L |
Estimated Shipment: 12-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only