Learn More
Thermo Scientific Chemicals Choline hydroxide, 45 wt.% aqueous solution, pure, stabilized
Brand: Thermo Scientific Chemicals 386720100
Code : 29
Additional Details : CAS Number : 123-41-1 Weight : 10.70000kg Transport : UN number : 3267 Chem class : 8 Pack group : II
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Choline hydroxide, 45 wt.% aqueous solution | |
White | |
Plastic drum | |
MFCD00002831 | |
1.07 | |
choline hydroxide, gossypine, sincaline, bursine, luridine, sinkalin, sinkaline, fagine, vidine, 2-hydroxy-n,n,n-trimethylethanaminium hydroxide | |
KIZQNNOULOCVDM-UHFFFAOYSA-M | |
2-hydroxyethyl(trimethyl)azanium;hydroxide | |
1800ppm ethylenediamine 3000ppm hydroxylamine | |
121.18 | |
Pure |
123-41-1,7732-18-5 | |
1.0700g/mL | |
C5H15NO2 | |
10 L | |
15,2211 | |
Solubility in water: miscible. | |
[OH-].C[N+](C)(C)CCO | |
121.18 | |
31255 | |
44.0 to 48.0% (Total base) | |
Granular Powder |
RUO – Research Use Only