Learn More
Christeyns™ Chriox Spray
Chriox spray is an ecological, ready-to-use disinfectant based on peracetic acid.
Brand:  Christeyns™ 391173
Additional Details : CAS Number : 64-19-7 Weight : 10.09000kg
 View more versions of this product
Product Code. 17800265
  • £81.30 / 5 L
Estimated Shipment: 28-08-2024
to see stock.
Add to basket
Description
Description
  • Eco-detergent certified by ECOCERT Greenlife according to the ECOCERT standard
  • Suitable for cleaning all washable surfaces (including in the food industry)
  • Spray the pure product on the surface to be treated or preferably on a clean, nonwoven cloth, and leave on for 5 to 15 min.
Specifications
Specifications
Surface Disinfectant
Clear Liquid
Unscented
2 Cans
No
Neutral
Non-sterile
5 L