missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ C18 Glass Column

Brand:  Macherey-Nagel™ 730004G

 View more versions of this product

Product Code. 10526701

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

Silica
6 mL
Solid Phase Extraction
Chromabond C18
Polar secondary interaction
500 m2/g
14%
SPE
45 μm
Solid Phase Extraction Column
60 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers