Learn More
Macherey-Nagel™ Chromabond™ HR-P Polypropylene SPE Column
Brand:  Macherey-Nagel™ 730118
 View more versions of this product
Product Code. 10338501
  • £201.00 / Pack of 30
Estimated Shipment: 21-11-2024
to see stock.
Explore more special offers
Add to basket
Specifications
6 mL
1 g
50 to 100 μm
Solid Phase Extraction Column
SPE
Solid Phase Extraction
Chromabond HR-P
1200 m2/g