missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Chromabond™ HR-P Polypropylene SPE Column

Brand:  Macherey-Nagel™ 730280

 View more versions of this product

Product Code. 10679001

  • £80.60 / Pack of 30
Estimated Shipment: 06-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Specifications

Specifications

1 mL
100 mg
Chromabond HR-P
1200 m2/g
SPE
50 to 100 μm
Solid Phase Extraction Column
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers